Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025670.4 | NP_079946.2 | 6 | 2829 | ||
GenBank mRNA | AK031705.1 | - | 6 | 2831 |