Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025677.3 | NP_079953.2 | 5 | 996 | ||
GenBank mRNA | AK004261.1 | - | 5 | 932 | ||
AK011608.1 | - | 5 | 953 | |||
AK017650.1 | - | 5 | 997 | |||
AK169429.1 | - | 4 | 3751 | |||
BC024342.1 | - | 5 | 926 | 5002659 |