Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nepn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025684.2 | NP_079960.1 | 2 | 936 | ||
GenBank mRNA | AK017656.1 | - | 2 | 937 | ||
AK017780.1 | - | 2 | 937 | |||
AK017842.1 | - | 2 | 806 | |||
AY938536.1 | - | 2 | 937 | |||
BC132046.1 | - | 2 | 926 | 40130323 | ||
BC132048.1 | - | 2 | 926 | 40130325 |