Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sltm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025690.3 | NP_079966.2 | 14 | 1888 | ||
NM_026337.1 | NP_080613.1 | 14 | 1926 | |||
GenBank mRNA | AF462146.1 | - | 14 | 1888 | ||
AK045723.1 | - | 8 | 988 | |||
AK134756.1 | - | 5 | 567 | |||
BC019992.1 | - | 3 | 258 | |||
BC139202.1 | - | 14 | 1878 | 8862224 | ||
BC145577.1 | - | 14 | 1878 | |||
CD806976.1 | - | 2 | 129 | |||
CN723089.1 | - | 3 | 345 |