Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcbld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025705.3 | NP_079981.2 | 7, 8 | 873 | ||
GenBank mRNA | AK014521.1 | - | 7, 8 | 685 | ||
AK016485.1 | - | 7, 8 | 873 | |||
BC026771.1 | - | 6, 7 | 661 | 4014778 |