Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gapvd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025709.2 | NP_079985.2 | 27 | 5803 | ||
GenBank mRNA | AK014474.1 | - | 12 | 2668 | ||
AK033450.1 | - | 77 | ||||
AK088851.1 | - | 11 | 2366 | |||
AK129381.1 | - | 26 | 5398 | |||
BC043715.1 | - | 6 | 1767 | |||
BC057164.1 | - | 12 | 2672 | |||
BC060123.1 | - | 27 | 5803 | 6408778 |