Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap3-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025720.3 | NP_079996.2 | 1 | 562 | ||
GenBank mRNA | AK003615.1 | - | 1 | 564 | ||
AK014641.1 | - | 1 | 563 | |||
BY703877.1 | - | 1 | 564 |