Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933434E20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287087.1 | NP_001274016.1 | 5 | 608 | ||
NM_025762.3 | NP_080038.1 | 3 | 463 | |||
NM_027500.4 | NP_081776.2 | 3 | 463 | |||
GenBank mRNA | AK017833.1 | - | 3 | 431 | ||
AK034749.1 | - | 3 | 426 | |||
AK051238.1 | - | 3 | 356 | |||
AK077215.1 | - | 5 | 609 | |||
AK150324.1 | - | 3 | 414 | |||
AK150423.1 | - | 3 | 414 | |||
AK150831.1 | - | 3 | 414 | |||
AK151002.1 | - | 3 | 413 | |||
AK151263.1 | - | 3 | 412 | |||
AK214769.1 | - | 3 | 213 | |||
AY533135.1 | - | 3 | 211 | |||
BC027190.1 | - | 3 | 323 | 3257131 | ||
BG091444.1 | - | 1 | 58 |