Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntnap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004357.2 | NP_001004357.2 | 23 | 3982 | ||
NM_025771.3 | NP_080047.1 | 3 | 429 | |||
GenBank mRNA | AK013139.1 | - | 3 | 397 | ||
AK017341.1 | - | 3 | 424 | |||
AK129232.1 | - | 23 | 3955 | |||
BC029826.1 | - | 3 | 378 | |||
BC064467.1 | - | 18 | 2873 | |||
BC103799.1 | - | 3 | 336 | |||
BC132074.1 | - | 2 | 241 | |||
BC132076.1 | - | 2 | 241 |