Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282485.1 | NP_001269414.1 | 2 | 290 | ||
NM_006259.2 | NP_006250.1 | 2 | 290 | |||
XM_005263126.3 | XP_005263183.1 | 2 | 725 | |||
XM_017008413.1 | XP_016863902.1 | 2 | 727 | |||
XM_017008414.1 | XP_016863903.1 | 2 | 722 | |||
XM_017008415.1 | XP_016863904.1 | 2 | 132 | |||
XM_017008416.1 | XP_016863905.1 | 2 | 644 | |||
XM_017008417.1 | XP_016863906.1 | 1 | 108 | |||
XM_017008418.1 | XP_016863907.1 | 1 | 108 | |||
GenBank mRNA | AK297673.1 | - | 2 | 237 | ||
D70899.1 | - | 1 | 95 | |||
DB455792.2 | - | 2 | 290 | |||
X94612.1 | - | 1 | 109 |