Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025785.2 | NP_080061.1 | 2 | 178 | ||
GenBank mRNA | AK008027.1 | - | 2 | 182 | ||
AK013645.1 | - | 2 | 179 | |||
AK018626.1 | - | 2 | 179 | |||
AK134722.1 | - | 2 | 182 | |||
BC014749.1 | - | 2 | 164 | 4240953 |