Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025786.3 | NP_080062.4 | 1 | 618 | ||
GenBank mRNA | AK020617.1 | - | 1 | 527 | ||
AK078919.1 | - | 1 | 613 | |||
BC011492.1 | - | 1 | 583 | 4207836 |