Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000112.3 | NP_000103.2 | 2 | 619 | ||
XM_017009191.1 | XP_016864680.1 | 2 | 627 | |||
GenBank mRNA | AK290358.1 | - | 2 | 442 | ||
AK312596.1 | - | 2 | 570 | |||
BC059390.1 | - | 2 | 473 | 30341764 | ||
U14528.1 | - | 1 | 378 |