Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tax1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025816.3 | NP_080092.2 | 2, 3 | 304 | ||
GenBank mRNA | AK004574.1 | - | 2, 3 | 286 | ||
AK146076.1 | - | 2, 3 | 284 | |||
AK171136.1 | - | 2, 3 | 271 | |||
BC014798.1 | - | 2, 3 | 225 | 3962810 |