Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1200014J11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025818.3 | NP_080094.3 | 2 | 251 | ||
GenBank mRNA | AK004766.1 | - | 2 | 239 | ||
AK011450.1 | - | 2 | 226 | |||
AK020130.1 | - | 2 | 247 | |||
AK033682.1 | - | 2 | 240 | |||
AK153152.1 | - | 2 | 239 | |||
AK153241.1 | - | 2 | 239 | |||
AK163780.1 | - | 2 | 248 | |||
AK165317.1 | - | 2 | 246 | |||
BC098495.1 | - | 2 | 223 | 30021633 | ||
BC118943.2 | - | 2 | 239 | 40092337 | ||
CJ093775.1 | - | 2 | 337 |