Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025832.2 | NP_080108.1 | 3, 4 | 294 | ||
GenBank mRNA | AK005056.1 | - | 3, 4 | 304 | ||
AK028432.1 | - | 3, 4 | 404 | |||
AK152498.1 | - | 3, 4 | 304 | |||
BC052445.1 | - | 3, 4 | 294 | 6412745 |