Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025839.3 | NP_080115.1 | 3 | 491 | ||
GenBank mRNA | AF435792.1 | - | 3 | 503 | ||
AK002378.1 | - | 3 | 484 | |||
AK003241.1 | - | 3 | 492 | |||
AK007897.1 | - | 3 | 487 | |||
AK159841.1 | - | 3 | 484 | |||
AK186853.1 | - | 2 | 201 | |||
BC025569.1 | - | 3 | 565 | 5322456 | ||
BC028494.1 | - | 3 | 484 | 1511165 |