Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025853.3 | NP_080129.2 | 3 | 227 | ||
GenBank mRNA | AK006246.1 | - | 3 | 228 | ||
AK017740.1 | - | 3 | 228 | |||
AK017813.1 | - | 3 | 182 | |||
AK145373.1 | - | 3 | 182 | |||
AK214124.1 | - | 2 | 122 | |||
BC025079.1 | - | 3 | 210 | 3499844 | ||
BC046807.1 | - | 3 | 188 | 6829342 |