Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284332.1 | NP_001271261.1 | 6 | 1072 | ||
NM_025858.3 | NP_080134.1 | 6 | 1069 | |||
NM_026381.5 | NP_080657.1 | 4 | 769 | |||
GenBank mRNA | AF520699.1 | - | 6 | 1050 | ||
AK009225.1 | - | 6 | 1009 | |||
AK018291.1 | - | 4 | 725 | |||
AK033545.1 | - | 6 | 1029 | |||
AK081060.1 | - | 4 | 747 | |||
AK088188.1 | - | 5 | 765 | |||
AK090357.1 | - | 4 | 923 | |||
AK090373.1 | - | 6 | 1008 | |||
AK150460.1 | - | 5 | 1227 | |||
AK151632.1 | - | 5 | 1227 | |||
AK183004.1 | - | 2 | 321 | |||
BC057089.1 | - | 4 | 721 | 5689110 |