Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025866.3 | NP_080142.1 | 3 | 289 | ||
GenBank mRNA | AK011289.1 | - | 3 | 290 | ||
AK028671.1 | - | 3 | 300 | |||
AK075882.1 | - | 3 | 254 | |||
AK153943.1 | - | 3 | 265 | |||
AK167734.1 | - | 3 | 290 | |||
BC066169.1 | - | 3 | 297 | 30474357 |