Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025789.5 | NP_080065.4 | 4, 5 | 1069 | ||
GenBank mRNA | AK007979.1 | - | 4, 5 | 702 | ||
AK016364.1 | - | 4, 5 | 655 | |||
AK148230.1 | - | 4, 5 | 1031 | |||
BC054439.1 | - | 4, 5 | 737 | |||
BC057649.1 | - | 4, 5 | 1028 | |||
BC085294.1 | - | 4, 5 | 717 |
Gene Symbol: Rsph3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083945.1 | NP_001077414.1 | 4, 5 | 758 | ||
GenBank mRNA | AK006083.1 | - | 4, 5 | 758 | ||
AK076872.1 | - | 4, 5 | 760 | |||
AK161285.1 | - | 4, 5 | 721 |