Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025799.4 | NP_080075.2 | 2 | 496 | ||
GenBank mRNA | AK002675.1 | - | 2 | 438 | ||
AK008653.1 | - | 2 | 441 | |||
AK017443.1 | - | 2 | 467 | |||
AK030320.1 | - | 2 | 498 | |||
AK035607.1 | - | 2 | 377 | |||
AK044790.1 | - | 2 | 353 | |||
AK146295.1 | - | 2 | 425 | |||
AK151204.1 | - | 2 | 357 | |||
AK159288.1 | - | 2 | 442 | |||
AK163042.1 | - | 2 | 440 | |||
BC004039.1 | - | 2 | 447 | 3496187 | ||
BC018181.1 | - | 2 | 456 | 3983067 | ||
BC094449.1 | - | 2 | 411 | 4222249 |