Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1200014J11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025818.3 | NP_080094.3 | 4 | 407 | ||
GenBank mRNA | AK004766.1 | - | 4 | 395 | ||
AK011450.1 | - | 4 | 382 | |||
AK017496.1 | - | 4 | 402 | |||
AK020130.1 | - | 4 | 403 | |||
AK032983.1 | - | 2 | 114 | |||
AK033682.1 | - | 4 | 396 | |||
AK153152.1 | - | 4 | 395 | |||
AK153241.1 | - | 4 | 395 | |||
AK166563.1 | - | 2 | 197 | |||
BC053720.1 | - | 2 | 196 | |||
BC098495.1 | - | 4 | 379 | 30021633 | ||
BC118943.2 | - | 4 | 395 | 40092337 | ||
CJ093775.1 | - | 4 | 493 |