Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025829.4 | NP_080105.1 | 4 | 327 | ||
GenBank mRNA | AK005054.2 | - | 4 | 328 | ||
AK007268.1 | - | 4 | 316 | |||
AY628329.1 | - | 4 | 308 | |||
BC027014.1 | - | 4 | 322 | 5369891 |