Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baiap2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025833.3 | NP_080109.1 | 5 | 573 | ||
GenBank mRNA | AK004918.2 | - | 5 | 574 | ||
AK008203.1 | - | 5 | 474 | |||
BC015459.1 | - | 5 | 351 | 3592632 |