Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000130.4 | NP_000121.2 | 3 | 468 | ||
XM_017000660.1 | XP_016856149.1 | 2 | 477 | |||
GenBank mRNA | AK226079.1 | - | 3 | 386 | ||
AK291613.1 | - | 3 | 418 | |||
AK300469.1 | - | 2 | 326 | |||
AU279657.1 | - | 3 | 468 | |||
M14335.1 | - | 3 | 399 | |||
M16967.1 | - | 3 | 413 |