Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025863.3 | NP_080139.3 | 3 | 521 | ||
GenBank mRNA | AK012269.1 | - | 3 | 522 | ||
AK016642.1 | - | 3 | 716 | |||
BC006700.1 | - | 3 | 439 | 3501285 | ||
BC025430.1 | - | 3 | 454 | 4017983 |