Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025768.2 | NP_080044.2 | 5, 6 | 674 | ||
GenBank mRNA | AF329833.1 | - | 3, 4 | 462 | ||
AK007905.1 | - | 5, 6 | 679 | |||
AK017197.1 | - | 5, 6 | 638 | |||
AK017251.1 | - | 6, 7 | 636 | |||
AK082863.1 | - | 5, 6 | 668 | |||
AK149322.1 | - | 671 | ||||
BC029096.1 | - | 5, 6 | 680 | 3500563 |