Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025783.3 | NP_080059.2 | 6 | 987 | ||
GenBank mRNA | AK009414.1 | - | 6 | 861 | ||
AK009447.1 | - | 1 | 232 | |||
AK018611.1 | - | 6 | 942 | |||
AK083562.1 | - | 6 | 988 | |||
BC027441.1 | - | 6 | 851 | |||
BC039759.1 | - | 6 | 862 | |||
BC049964.1 | - | 6 | 908 | 3154282 |