Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025785.2 | NP_080061.1 | 3 | 277 | ||
GenBank mRNA | AK008027.1 | - | 3 | 281 | ||
AK018626.1 | - | 3 | 278 | |||
AK134722.1 | - | 3 | 281 | |||
BC014749.1 | - | 3 | 263 | 4240953 |