Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163689.1 | NP_001157161.1 | 8 | 1261 | ||
NM_025802.3 | NP_080078.2 | 7 | 1093 | |||
GenBank mRNA | AK002826.1 | - | 7 | 1075 | ||
AK003207.1 | - | 3 | 315 | |||
AK031609.1 | - | 8 | 1219 | |||
AK076464.1 | - | 8 | 1045 | |||
AK085926.1 | - | 8 | 1246 | |||
AK150184.1 | - | 8 | 1243 | |||
AY510273.1 | - | 8 | 1242 | |||
AY731699.1 | - | 8 | 1242 | |||
AY894805.1 | - | 8 | 1162 | |||
BC019188.1 | - | 9 | 1325 | 5027544 | ||
BC034190.1 | - | 7 | 762 | |||
BC044737.1 | - | 9 | 1307 | |||
BC044781.2 | - | 9 | 1307 | 5151911 | ||
BC054825.1 | - | 9 | 1223 |