Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tax1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025816.3 | NP_080092.2 | 3, 4 | 407 | ||
GenBank mRNA | AK004574.1 | - | 3, 4 | 389 | ||
AK146076.1 | - | 3, 4 | 387 | |||
AK171136.1 | - | 3, 4 | 374 | |||
BC014798.1 | - | 3, 4 | 328 | 3962810 |