Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025824.3 | NP_080100.1 | 4, 5 | 435 | ||
GenBank mRNA | AK004784.2 | - | 4, 5 | 435 | ||
AK013980.1 | - | 4, 5 | 429 | |||
AK029030.1 | - | 4, 5 | 446 | |||
AK076086.1 | - | 4, 5 | 436 | |||
AK076560.1 | - | 4, 5 | 436 | |||
AK087980.1 | - | 4, 5 | 423 | |||
AK088322.1 | - | 4, 5 | 443 | |||
AK133973.1 | - | 4, 5 | 436 | |||
AK151919.1 | - | 4, 5 | 423 | |||
AK152939.1 | - | 4, 5 | 423 | |||
AK153538.1 | - | 4, 5 | 424 | |||
AK161192.1 | - | 4, 5 | 433 | |||
AK166247.1 | - | 4, 5 | 440 | |||
AK167056.1 | - | 4, 5 | 420 | |||
AK167074.1 | - | 4, 5 | 407 | |||
AK167514.1 | - | 4, 5 | 395 | |||
AK167982.1 | - | 4, 5 | 441 | |||
AK168158.1 | - | 4, 5 | 447 | |||
AK198677.1 | - | 1, 2 | 50 | |||
AK212098.1 | - | 4, 5 | 399 | |||
AK215707.1 | - | 23 | ||||
BC005466.1 | - | 4, 5 | 444 | 3601190 | ||
BC028865.1 | - | 4, 5 | 423 | 4910833 |