Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025826.4 | NP_080102.1 | 6, 7 | 960 | ||
GenBank mRNA | AK004889.1 | - | 6, 7 | 861 | ||
AK076535.1 | - | 6, 7 | 861 | |||
AK131628.1 | - | 6, 7 | 907 | |||
AK152325.1 | - | 6, 7 | 849 | |||
AK163322.1 | - | 2, 3 | 441 | |||
BC011235.1 | - | 6, 7 | 842 | |||
BC035231.1 | - | 6, 7 | 826 | |||
BC044826.1 | - | 6, 7 | 893 | |||
BC054428.1 | - | 6, 7 | 902 | 5039627 |