Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lman2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025828.3 | NP_080104.2 | 8 | 1698 | ||
GenBank mRNA | AK004952.1 | - | 8 | 1529 | ||
AK014384.1 | - | 8 | 1520 | |||
AK083442.1 | - | 8 | 1531 | |||
AK129028.1 | - | 8 | 1525 | |||
AK169401.1 | - | 8 | 1526 | |||
AK219874.1 | - | 1 | 103 | |||
BC055327.1 | - | 8 | 1698 | 5702182 |