Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025839.3 | NP_080115.1 | 3 | 1237 | ||
GenBank mRNA | AF435792.1 | - | 3 | 1249 | ||
AK002378.1 | - | 3 | 1230 | |||
AK003241.1 | - | 3 | 1238 | |||
AK007897.1 | - | 3 | 1233 | |||
AK019936.1 | - | 4 | 1125 | |||
AK159841.1 | - | 3 | 1230 | |||
AK182660.1 | - | 1 | 193 | |||
AK204706.1 | - | 1 | 193 | |||
AK205794.1 | - | 1 | 193 | |||
AK207255.1 | - | 1 | 193 | |||
AK213600.1 | - | 1 | 193 | |||
AK214569.1 | - | 1 | 193 | |||
AK216077.1 | - | 1 | 193 | |||
AK218747.1 | - | 1 | 193 | |||
BC025569.1 | - | 3 | 1311 | 5322456 | ||
BC028494.1 | - | 5 | 752 | 1511165 |