Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cir1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025854.3 | NP_080130.2 | 3, 4 | 341 | ||
GenBank mRNA | AK006260.1 | - | 3, 4 | 253 | ||
AK008242.1 | - | 3, 4 | 305 | |||
AK131674.1 | - | 3, 4 | 317 | |||
BC012219.1 | - | 3, 4 | 450 | |||
BC061077.1 | - | 3, 4 | 276 | |||
BC089602.1 | - | 3, 4 | 289 | |||
BC094283.1 | - | 3, 4 | 277 | |||
BC099444.1 | - | 3, 4 | 316 | |||
CJ050326.1 | - | 3, 4 | 342 |