Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284332.1 | NP_001271261.1 | 6 | 1677 | ||
NM_025858.3 | NP_080134.1 | 6 | 1674 | |||
NM_026381.5 | NP_080657.1 | 4 | 1374 | |||
GenBank mRNA | AF520699.1 | - | 6 | 1655 | ||
AK009225.1 | - | 6 | 1614 | |||
AK018291.1 | - | 4 | 1326 | |||
AK033545.1 | - | 6 | 1634 | |||
AK081060.1 | - | 4 | 1352 | |||
AK088188.1 | - | 5 | 1370 | |||
AK090357.1 | - | 4 | 1529 | |||
AK090373.1 | - | 6 | 1613 | |||
AK150460.1 | - | 5 | 1832 | |||
AK151632.1 | - | 5 | 1832 | |||
AK182311.1 | - | 1 | 343 | |||
AK207879.1 | - | 1 | 358 | |||
BC010238.1 | - | 6 | 1638 | 3581921 | ||
BC017600.1 | - | 4 | 1286 | 4487600 | ||
BC057089.1 | - | 4 | 1326 | 5689110 |