Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000147.4 | NP_000138.2 | 2, 3 | 615 | ||
XM_005245821.2 | XP_005245878.1 | 2, 3 | 752 | |||
XM_011541167.2 | XP_011539469.1 | 2, 3 | 200 | |||
XM_017000905.1 | XP_016856394.1 | 2, 3 | 297 | |||
GenBank mRNA | AB074175.1 | - | 2, 3 | 536 | ||
AK092914.1 | - | 3, 4 | 470 | |||
AK095443.1 | - | 3, 4 | 487 | |||
AK095455.1 | - | 3, 4 | 501 | |||
AK225468.1 | - | 2, 3 | 536 | |||
AK295535.1 | - | 2, 3 | 719 | |||
AK301302.1 | - | 3, 4 | 490 | |||
AK310143.1 | - | 3, 4 | 598 | |||
AK314649.1 | - | 2, 3 | 536 | |||
BC017338.2 | - | 2, 3 | 538 | 4871788 | ||
M10355.1 | - | 2, 3 | 310 | |||
M29877.1 | - | 2, 3 | 535 | |||
X01390.1 | - | 2, 3 | 310 |