Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025887.4 | NP_080163.1 | 2 | 602 | ||
GenBank mRNA | AB232593.1 | - | 1 | 135 | ||
AK002631.1 | - | 2 | 357 | |||
AK008198.1 | - | 2 | 343 | |||
AK010495.1 | - | 2 | 447 | |||
AK076093.1 | - | 2 | 583 | |||
AK081999.1 | - | 2 | 603 | |||
AK150296.1 | - | 2 | 356 | |||
AK150346.1 | - | 2 | 356 | |||
AK150405.1 | - | 2 | 400 | |||
AK151066.1 | - | 2 | 356 | |||
AK153131.1 | - | 2 | 356 | |||
AK159706.1 | - | 2 | 356 | |||
AK177306.1 | - | 368 | ||||
BC004842.1 | - | 2 | 463 | 2649317 | ||
BC034370.1 | - | 2 | 345 | 3709729 | ||
BC096481.1 | - | 3 | 536 | 5322132 |