Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025891.3 | NP_080167.3 | 4 | 541 | ||
GenBank mRNA | AK005094.1 | - | 4 | 488 | ||
AK158019.1 | - | 516 | ||||
BC013122.1 | - | 4 | 578 | 3586276 | ||
BC060525.1 | - | 4 | 440 | 6817219 |