Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025900.2 | NP_080176.2 | 2 | 277 | ||
GenBank mRNA | AK012315.1 | - | 2 | 232 | ||
AK012615.1 | - | 2 | 218 | |||
AK012877.1 | - | 2 | 212 | |||
AK033451.1 | - | 2 | 167 | |||
AK154013.1 | - | 2 | 259 | |||
AV449751.1 | - | 2 | 278 | |||
BC055451.1 | - | 2 | 223 | 4164972 | ||
BU920290.1 | - | 2 | 261 |