Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025901.3 | NP_080177.1 | 1, 2 | 218 | ||
GenBank mRNA | AK005152.1 | - | 1, 2 | 122 | ||
AK008637.1 | - | 1, 2 | 90 | |||
AK010875.1 | - | 1, 2 | 156 | |||
AK021353.1 | - | 1, 2 | 153 | |||
AK031110.1 | - | 1, 2 | 153 | |||
AK088206.1 | - | 1, 2 | 118 | |||
BC025898.1 | - | 1, 2 | 234 | |||
BY003788.1 | - | 1, 2 | 220 |