Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yae1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025904.3 | NP_080180.1 | 1, 2 | 182 | ||
GenBank mRNA | AK005425.1 | - | 1, 2 | 169 | ||
AK011774.1 | - | 1, 2 | 181 | |||
AK011805.1 | - | 1, 2 | 150 | |||
AK011852.1 | - | 1, 2 | 150 | |||
AK019184.1 | - | 1, 2 | 136 | |||
AK030455.1 | - | 1 | 1593 | |||
AK076034.1 | - | 1, 2 | 139 | |||
AK151981.1 | - | 1, 2 | 160 | |||
BC020123.1 | - | 2 | 176 | |||
BC037189.1 | - | 1, 2 | 169 | |||
BC048453.1 | - | 1, 2 | 165 | 6595060 | ||
BC083111.1 | - | 1, 2 | 141 | 5329860 | ||
CJ084672.1 | - | 1, 2 | 182 |