Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025907.3 | NP_080183.1 | 2 | 431 | ||
GenBank mRNA | AK005448.1 | - | 2 | 486 | ||
AK006715.1 | - | 2 | 473 | |||
AK078185.1 | - | 2 | 485 | |||
AK082949.1 | - | 2 | 486 | |||
AK133735.1 | - | 2 | 463 | |||
BC030449.1 | - | 2 | 432 | 5400506 |