Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277273.1 | NP_001264202.1 | 1 | 217 | ||
NM_025923.3 | NP_080199.1 | 1 | 217 | |||
GenBank mRNA | AF513619.1 | - | 1 | 217 | ||
AK008605.1 | - | 1 | 103 | |||
AK011520.1 | - | 1 | 122 | |||
AK011752.1 | - | 1 | 94 | |||
AK011903.1 | - | 1 | 100 | |||
AK017431.1 | - | 1 | 100 | |||
AK018018.1 | - | 1 | 99 | |||
AK020981.1 | - | 1 | 100 | |||
AK076017.1 | - | 1 | 100 | |||
AK168538.1 | - | 1 | 101 | |||
BC032876.1 | - | 1 | 91 | 1282316 | ||
BC066181.1 | - | 1 | 146 | 6435171 |