Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000152.4 | NP_000143.2 | 16 | 2604 | ||
NM_001079803.2 | NP_001073271.1 | 17 | 2419 | |||
NM_001079804.2 | NP_001073272.1 | 16 | 2339 | |||
XM_005257193.2 | XP_005257250.1 | 17 | 2405 | |||
XM_005257194.4 | XP_005257251.1 | 17 | 2445 | |||
GenBank mRNA | BC040431.1 | - | 17 | 2351 | 5739991 | |
DQ907243.1 | - | 15 | 2207 | |||
M34424.1 | - | 15 | 2239 | |||
Y00839.1 | - | 16 | 2426 |