Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025932.2 | NP_080208.2 | 1, 2 | 393 | ||
GenBank mRNA | AK008390.1 | - | 1, 2 | 362 | ||
AK011337.1 | - | 1, 2 | 393 | |||
AK014893.1 | - | 1, 2 | 379 | |||
AK051251.1 | - | 1, 2 | 350 | |||
AK147055.1 | - | 1, 2 | 329 | |||
AK154843.1 | - | 1, 2 | 350 | |||
AK161413.1 | - | 1, 2 | 350 | |||
BC021373.1 | - | 1, 2 | 323 | 5039534 |