Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291108.1 | NP_001278037.1 | 2, 3 | 413 | ||
NM_025947.3 | NP_080223.2 | 2, 3 | 169 | |||
GenBank mRNA | AK002359.1 | - | 2, 3 | 110 | ||
AK003435.1 | - | 1, 2 | 99 | |||
AK008214.1 | - | 2, 3 | 131 | |||
AK008595.1 | - | 3, 4 | 131 | |||
AK166685.1 | - | 2, 3 | 145 | |||
BC099423.1 | - | 2, 3 | 129 | 30933448 | ||
BC125610.1 | - | 2, 3 | 129 | 40130125 | ||
BC125648.1 | - | 2, 3 | 129 | 40130163 | ||
CJ136086.1 | - | 2, 3 | 414 |