Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025974.2 | NP_080250.1 | 4 | 267 | ||
GenBank mRNA | AK002946.1 | - | 4 | 268 | ||
AK007807.1 | - | 4 | 268 | |||
AK010600.1 | - | 4 | 270 | |||
AK012277.1 | - | 4 | 268 | |||
AK013912.1 | - | 4 | 268 | |||
AK141887.1 | - | 3 | 825 | |||
AK167146.1 | - | 4 | 268 | |||
BC013249.1 | - | 1 | 26 | |||
BC013251.1 | - | 1 | 26 | |||
BC037202.1 | - | 3 | 268 | |||
BC042937.1 | - | 3 | 246 |
Gene Symbol: Rpl14-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC008177.1 | - | 1 | 116 | ||
BC092249.1 | - | 1 | 252 | 5034997 |